Learn More
Medchemexpress LLC Citric acid-d4 | 147664-83-3 | MFCD00209702 | 99.3% | 196.15 | C6H4D4O7 | 10 MG
SDP
Catalog No. 5000296466
Encompass_Preferred
Citric acid-d4 is the deuterium-labeled form of citric acid supplied as a white to off-white solid. It serves as a stable-isotope reference standard for analytical workflows, including mass spectrometry, NMR, isotopic tracer experiments, and quantitative assays. Molecular formula C6H4D4O7; molecular weight 196.15; purity 99.31%.
  • Deuterium labeled (d4) for isotope dilution and tracer studies.
  • High purity for reliable quantitation.
  • Solid form simplifies weighing and solution preparation.
  • Available in small research-sized pack quantities.
  • Stable when stored sealed away from moisture and light.
Catalog No. 50-002-96466 Supplier Medchemexpress LLC Supplier No. HYN1428S10MG
Citric acid-d4 is the deuterium-labeled form of citric acid supplied as a white to off-white solid. It serves as a stable-isotope reference standard for analytical workflows, including mass spectrometry, NMR, isotopic tracer experiments, and quantitative assays. Molecular formula C6H4D4O7; molecular weight 196.15; purity 99.31%.
  • Deuterium labeled (d4) for isotope dilution and tracer studies.
  • High purity for reliable quantitation.
  • Solid form simplifies weighing and solution preparation.
  • Available in small research-sized pack quantities.
  • Stable when stored sealed away from moisture and light.