Alfa Aesar™ Citric acid diammonium salt, ACS, 98.0-103.0%

Manufacturer: Alfa Aesar™ 36219A3

Quantity 2kg

 View more versions of this product

Catalog No. AA36219A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3012-65-5
    Formula Weight 226.19
    MDL Number MFCD00013068
    Merck Index 14,512
    Synonym Ammonium hydrogen citrate; Ammonium citrate dibasic
    Chemical Name or Material Citric acid diammonium salt
    Grade ACS Grade

    Specifications

    Formula Weight 226.19
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Granular
    CAS 3012-65-5
    Synonym Ammonium hydrogen citrate; Ammonium citrate dibasic
    Health Hazard 1 Warning
    Quantity 2kg
    Merck Index 14,512
    Density 1.48g/mL
    Odor Odorless
    TSCA TSCA
    MDL Number MFCD00013068
    Chemical Name or Material Citric acid diammonium salt
    Assay 98.0 to 103.0%
    Grade ACS Grade

    GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit