Learn More
MSC CITRIC ACID DISINFECTANT
Catalog No. 502815638
Encompass
Contec Citric Acid Disinfectant is a gentle, low-odor botanical formula, ideal for restoration jobs and environments with chemical sensitivities. This highly effective disinfectant eliminates viruses, bacteria, fungi, and tuberculosis.
  • Eliminates viruses, bacteria, fungi, and tuberculosis in 5–10 minutes
  • Combats tough non-enveloped viruses including Norovirus, Poliovirus, and SARS-CoV-2
  • Broad-spectrum solution for challenging cleaning tasks
  • EPA List N product (EPA Registration Number: 34810-35)
Catalog No. 50-281-5638 Supplier MSC Supplier No. 11571064
Contec Citric Acid Disinfectant is a gentle, low-odor botanical formula, ideal for restoration jobs and environments with chemical sensitivities. This highly effective disinfectant eliminates viruses, bacteria, fungi, and tuberculosis.
  • Eliminates viruses, bacteria, fungi, and tuberculosis in 5–10 minutes
  • Combats tough non-enveloped viruses including Norovirus, Poliovirus, and SARS-CoV-2
  • Broad-spectrum solution for challenging cleaning tasks
  • EPA List N product (EPA Registration Number: 34810-35)