Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, disodium salt sesquihydrate, 99%, pure

Catalog No. AC250240025
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250240025 2.5 kg Plastic Bottle
AC250242500 250 g Plastic Bottle
AC250240010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC250240025 Supplier Thermo Scientific Chemicals Supplier No. 250240025
Add to Cart

CAS: 1545832 | C12H18Na4O17 | 526.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1545832
Molecular Formula C12H18Na4O17
Molecular Weight (g/mol) 526.22
MDL Number MFCD00150445
InChI Key HGPVLOQNBSHYEI-UHFFFAOYNA-J
Synonym unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3
PubChem CID 117063816
IUPAC Name tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate
SMILES C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 149.0°C
Color White
pH 4.9 to 5.2 (3% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic Bottle
Linear Formula 1·5H2O
Quantity 2.5 kg
Merck Index 14, 2326
Solubility Information Solubility in water: 500g/L (20°C).
Formula Weight 263.1
Percent Purity 0.99
Grade Pure
Physical Form Powder
Chemical Name or Material Citric acid, disodium salt sesquihydrate, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.