Learn More
Hach Company Citric Acid Reagent Powder Pillows for Silica, 10 ml, 100/pk
Catalog No. 50465271 Shop All Hach Company Products
Encompass
For determination of high range silica by the silicomolybdate method (Hach method 8185), and low range silica by the heteropoly blue method (Hach method 8186). 100/pk
Catalog No. 50-465-271 Supplier Hach Company Supplier No. 2106269
For determination of high range silica by the silicomolybdate method (Hach method 8185), and low range silica by the heteropoly blue method (Hach method 8186). 100/pk