Learn More
Selleck Chemical LLC Citric acid monohydrate E2164-100MG
SDP
Catalog No. 5000039152
Encompass_Preferred
Citric acid monohydrate is a natural preservative and food tartness enhancer Citric acid monohydrate induces apoptosis and cell cycle arrest at G2/M phase and S phase Citric acid monohydrate cause oxidative damage of the liver by means of the decrease of antioxidative enzyme activities Citric acid monohydrate causes renal toxicity in mice
Catalog No. 50-000-39152 Supplier Selleck Chemical LLC Supplier No. E2164100MG
Citric acid monohydrate is a natural preservative and food tartness enhancer Citric acid monohydrate induces apoptosis and cell cycle arrest at G2/M phase and S phase Citric acid monohydrate cause oxidative damage of the liver by means of the decrease of antioxidative enzyme activities Citric acid monohydrate causes renal toxicity in mice