Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Citric Acid Monohydrate

Catalog No. C7129100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C7129100G 100 g
C71291KG 1 kg
C7129500G 500 g
C71295KG 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. C7129100G Supplier Sigma Aldrich Supplier No. C7129100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Citric acid monohydrate(CAM) is an organic acid. It can be produced by crystallization from mother liquor of citric acid solution at 20-25C during citric acid synthesis. Its molar enthalpy of solution in water has been reported to be +÷solHm (298.15K, m = 0.0203molkg-1) = (29061+/-123)Jmol-1. An investigation of its crystal growth kinetics indicates that growth is linearly dependent on size.Citric acid monohydrate may be used:To prepare citric acid solution employed in the acetone method of 68Ga pre-purification and radiolabeling technique.To prepare Tris-citrate buffer employed for the electrophoresis of bacterial enzymes.To improve the release of diltiazem hydrochloride from melt extruded matrix tablets.

Specifications

CAS 5949-29-1
Quantity 100 g
Molecular Formula C6H8O7 -+ H2O
Linear Formula HOC(COOH)(CH2COOH)2 -+ H2O
MDL Number MFCD00149972
Molecular Weight (g/mol) 210.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.