Learn More
Supply Solutions Citric acid monohydrate, 5949-29-1, MFCD00149972, 500g
Catalog No. NC1593448
Encompass_Preferred
Molecular Formula C6H8O7.H2O, Purity ?99.0%, Molecular Weight 210.137
Catalog No. NC1593448 Supplier Supply Solutions Supplier No. C1909500G
Molecular Formula C6H8O7.H2O, Purity ?99.0%, Molecular Weight 210.137