Learn More
Medchemexpress LLC Citric acid monohydrate | 5949-29-1 | MFCD00149972 | 210.14 | 100 G
SDP
Catalog No. 5000415324
Encompass_Preferred
Citric acid monohydrate is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. It can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries. This product is for research use only and not sold to patients.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest
  • Acidulant
  • Emulsifier
  • Sequestrant
  • Buffering agent
Catalog No. 50-004-15324 Supplier Medchemexpress LLC Supplier No. HYN1428A100G
Citric acid monohydrate is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. It can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries. This product is for research use only and not sold to patients.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest
  • Acidulant
  • Emulsifier
  • Sequestrant
  • Buffering agent