Learn More
Alkali Scientific Citric Acid, Monohydrate, ACS Grade, 5 Kg
SDP
Catalog No. 5000659896
Encompass_Preferred
Citric Acid Monohydrate, ACS Grade, 5 kg, is a high-purity reagent used in a wide range of laboratory and industrial applications. Citric acid is central to energy metabolism and acts as a natural preservative in food and pharmaceuticals. Its role in buffer systems, as a pH adjuster, and in the chelation of metal ions makes it a versatile reagent in scientific research. The 5 kg package is well-suited for labs and manufacturing processes that require large quantities of citric acid, such as biochemistry experiments, industrial scale synthesis, and production of food and cosmetic products. Its high purity ensures reliable results in sensitive experiments.
Catalog No. 50-006-59896 Supplier Alkali Scientific Supplier No. ITR007CP
Citric Acid Monohydrate, ACS Grade, 5 kg, is a high-purity reagent used in a wide range of laboratory and industrial applications. Citric acid is central to energy metabolism and acts as a natural preservative in food and pharmaceuticals. Its role in buffer systems, as a pH adjuster, and in the chelation of metal ions makes it a versatile reagent in scientific research. The 5 kg package is well-suited for labs and manufacturing processes that require large quantities of citric acid, such as biochemistry experiments, industrial scale synthesis, and production of food and cosmetic products. Its high purity ensures reliable results in sensitive experiments.