Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid Monohydrate Bioreagent Grade, Vion Biosciences
SCI_ED

Catalog No. S11080 Shop All Aldon Products
Change view
Click to view available options
Quantity:
10 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11080 25 kg
S11072 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
WARNING: Causes serious eye irritation.
Catalog No. S11080 Supplier Aldon Corporation Supplier No. VNCC700325KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: Causes serious eye irritation.

Bioreagent grade for molecular biology, DNase-free, Rnase-free, ≥ 99%

  • SPECIFICATIONS
  • Appearance (Color): Colorless or White
  • Appearance (Form): Crystals or Crystalline Powder
  • Purity: 99.0 - 102.0%
  • Identification (FTIR): Conforms to Structure
  • Water (KFV): 7.5 - 8.8%
  • Lead: ≤ 3 ppm
  • Iron: ≤ 3 ppm
  • Oxalate: To Pass Test
  • Sulfate: ≤ 20 ppm
  • Chloride: ≤ 10 ppm
  • Insoluble Matter: ≤ 0.005%
  • Residue on Ignition: ≤ 0.02%
  • DNase: None Detected
  • RNase: None Detected

Specifications

Quantity 25 kg
CAS 77-92-9
Packaging UN Plastic Pail
Molecular Weight (g/mol) 192.13 g/mol
Percent Purity 99.0 to 102.0%
Grade Reagent
Chemical Name or Material Citric acid, anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.