Learn More
Citric Acid Monohydrate, MP Biomedicals™
Catalog No. MP021506995 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
MP021506995 500 g
1 options
Catalog No. MP021506995 Supplier MP Biomedicals, Inc Supplier No. 02150699.5
Add to Cart
Add to Cart
Acts as an iron chelator. Citric Acid Monohydrate, MP Biomedicals is an organic acid that can be produced by crystallization from mother liquor at 20–25deg;C during citric acid synthesis. It is a useful buffer component and can support biosynthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
Specifications
Quantity 500 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov