Learn More
Citric Acid Monohydrate, TraceSELECT™, ≥99.9998% (Metals basis), Solstice
Shop All Solstice Research Chemicals ProductsDescription
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Specifications
| pH | 1.8 (at 50.00g/L 20°C) |
| Melting Point | ∼135°C to 152°C |
| Color | White |
| Density | ∽1.54g/cu.cm at 20°C |
| Quantity | 100 g |
| Flash Point | 173°C (343.4°F) |
| Odor | Odorless |
| Packaging | Poly bottle |
| Physical Form | Crystalline |
| Linear Formula | HOC(COOH)(CH2COOH)2 · H2O |
| Show More |
Safety and Handling
| Hazard Statement | P305 + P351 + P338 |
| RTECSNumber | 201-069-1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.