Learn More
LAB ALLEY Lab Alley Citric Acid Granular USP/FCC/Food Grade Kosher Halal
SDP
Catalog No. NC2291050
Encompass
Citric Acid Granular USP/NF/FCC/Food Grade Kosher from Lab Alley is a compound that meets all the requirements of the United States Pharmacopeia (USP) acceptable for food drug or medicinal use. Chemicals Codex (FCC) that refers to a compendium of standards that is used internationally to ensure the quality and purity of food ingredients. This product is also regulated by the Food and Drug Administration and is internationally recognized for its identity purity and quality as a food product. Citric Acid from Lab Alley is colorless to white translucent granular with no odor and thanks to its high level of purity it can be generally used in the food industry as an acidifier flavoring agent and chelating agent as well as in the pharmaceutical industry. Citric Acid Granular USP/FCC/Food Grade Kosher is a kitchen essential commonly utilized for preserving flavoring and preserving the Vitamin C content of a food product.
This item is not returnable. View return policy
Catalog No. NC2291050 Supplier LAB ALLEY Supplier No. CTAGF1LB
Citric Acid Granular USP/NF/FCC/Food Grade Kosher from Lab Alley is a compound that meets all the requirements of the United States Pharmacopeia (USP) acceptable for food drug or medicinal use. Chemicals Codex (FCC) that refers to a compendium of standards that is used internationally to ensure the quality and purity of food ingredients. This product is also regulated by the Food and Drug Administration and is internationally recognized for its identity purity and quality as a food product. Citric Acid from Lab Alley is colorless to white translucent granular with no odor and thanks to its high level of purity it can be generally used in the food industry as an acidifier flavoring agent and chelating agent as well as in the pharmaceutical industry. Citric Acid Granular USP/FCC/Food Grade Kosher is a kitchen essential commonly utilized for preserving flavoring and preserving the Vitamin C content of a food product.