Learn More
Hach Company Citric Acid Powder Pillows for Silica, 25 mL, pk/1000
Supplier:  Hach Company 1454828
Encompass
For silica determination by photometric methods. Sample Size: 25 mL Pack of 100 powder pillows.
Catalog No. 50-465-270