Learn More
Hach Company Citric Acid F Reagent Solution, 500ml
Supplier:  Hach Company 2254249
Encompass
For determination of ultra-low range silica by the Heteropoly Blue method (Hach Method 8282). Range: 3-1000 µg/l SiO2. 500ml
Catalog No. 50-465-269