Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid Solution, 50% by weight, Biotech Reagent, J.T. Baker™

Catalog No. 02002624 Shop All Avantor Products
Change view
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-624 19 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-624 Supplier Avantor Supplier No. 034107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
IUPAC Name ammonia
SMILES N

Specifications

Quantity 19 L
Packaging Poly Bottle
Specific Gravity 1L = 1.24kg
Formula Weight 192.12
Percent Purity 50%
Grade Biotech Reagent
Chemical Name or Material Citric Acid Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.