Learn More
Medchemexpress LLC Citric acid (standard) | 77-92-9 | 192.12 | 25 MG
SDP
Catalog No. 5000389881
Encompass_Preferred
Citric acid (standard) is an analytical standard for research and analytical applications. It acts as a natural preservative and food tartness enhancer, also serving as an acidulant, emulsifier, sequestrant, and buffering agent. Research indicates its role in inducing apoptosis and cell cycle arrest in HaCaT cells and its potential to cause oxidative liver damage.
  • Analytical standard for research and analytical applications
  • Natural preservative and food tartness enhancer
  • Induces apoptosis and cell cycle arrest in HaCaT cells
  • Can cause oxidative damage of the liver
  • Functions as an acidulant, emulsifier, sequestrant, and buffering agent
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-89881 Supplier Medchemexpress LLC Supplier No. 25MGHYN1428R
Citric acid (standard) is an analytical standard for research and analytical applications. It acts as a natural preservative and food tartness enhancer, also serving as an acidulant, emulsifier, sequestrant, and buffering agent. Research indicates its role in inducing apoptosis and cell cycle arrest in HaCaT cells and its potential to cause oxidative liver damage.
  • Analytical standard for research and analytical applications
  • Natural preservative and food tartness enhancer
  • Induces apoptosis and cell cycle arrest in HaCaT cells
  • Can cause oxidative damage of the liver
  • Functions as an acidulant, emulsifier, sequestrant, and buffering agent
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)