Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citric acid (standard) | 77-92-9 | MFCD00011669 | 99.9% | 192.12 g/mol | C6H8O7 | 10 MG
SDP

Catalog No. 5000295707
Encompass_Preferred

Analytical citric acid standard for laboratory calibration and method development. Supplied as a small, high-purity reference material with accompanying documentation for analytical use.

  • High purity: 99.88%.
  • Molecular formula: C6H8O7.
  • CAS number: 77-92-9.
  • Supplied in small, precise quantities suitable for analytical workflows.
  • Includes certificate of analysis and safety data sheet for traceability.

Catalog No. 50-002-95707 Supplier Medchemexpress LLC Supplier No. HYN1428R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical citric acid standard for laboratory calibration and method development. Supplied as a small, high-purity reference material with accompanying documentation for analytical use.

  • High purity: 99.88%.
  • Molecular formula: C6H8O7.
  • CAS number: 77-92-9.
  • Supplied in small, precise quantities suitable for analytical workflows.
  • Includes certificate of analysis and safety data sheet for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.