Learn More
Medchemexpress LLC Citric acid triammonium | 3458-72-8 | 99.87% | 5 G
SDP
Catalog No. 5000411380
Encompass_Preferred
Citric acid triammonium is formed by Citric acid reacting with ammonia in a molar ratio of 1:3. It can be utilized as a carbon source for preparing carbon quantum dots (CDs). The presence of higher nitrogen components may enhance the nitrogen-based functional groups in CDs, resulting in more efficient emission-color tunability. This product is for research use only and not for sale to patients.
  • Formed by reaction of citric acid with ammonia in 1:3 molar ratio
  • Utilized as a carbon source for preparing carbon quantum dots (CDs)
  • May promote nitrogen-based functional groups in CDs
  • Leads to more efficient emission-color tunability in CDs
Catalog No. 50-004-11380 Supplier Medchemexpress LLC Supplier No. HYB1529A5G
Citric acid triammonium is formed by Citric acid reacting with ammonia in a molar ratio of 1:3. It can be utilized as a carbon source for preparing carbon quantum dots (CDs). The presence of higher nitrogen components may enhance the nitrogen-based functional groups in CDs, resulting in more efficient emission-color tunability. This product is for research use only and not for sale to patients.
  • Formed by reaction of citric acid with ammonia in 1:3 molar ratio
  • Utilized as a carbon source for preparing carbon quantum dots (CDs)
  • May promote nitrogen-based functional groups in CDs
  • Leads to more efficient emission-color tunability in CDs