Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citric acid triammonium | 3458-72-8 | 99.9% | 100 G
SDP

Catalog No. 5000411938
Encompass_Preferred

Citric acid triammonium is formed by Citric acid reacting with ammonia in a molar ratio of 1:3. It can be utilized as a carbon source for preparing carbon quantum dots (CDs). When citric acid triammonium contains higher nitrogen components, it may enhance the nitrogen-based functional groups in CDs, leading to more efficient emission-color tunability.

  • Molecular weight: 243.22
  • Formula: C6H17N3O7
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C(CC(C(O)=O)(O)CC(O)=O)O.[3NH3]
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Catalog No. 50-004-11938 Supplier Medchemexpress LLC Supplier No. HYB1529A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citric acid triammonium is formed by Citric acid reacting with ammonia in a molar ratio of 1:3. It can be utilized as a carbon source for preparing carbon quantum dots (CDs). When citric acid triammonium contains higher nitrogen components, it may enhance the nitrogen-based functional groups in CDs, leading to more efficient emission-color tunability.

  • Molecular weight: 243.22
  • Formula: C6H17N3O7
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C(CC(C(O)=O)(O)CC(O)=O)O.[3NH3]
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.