Learn More
Medchemexpress LLC Citric acid triammonium | 3458-72-8 | 99.9% | 50 G
SDP
Catalog No. 5000411197
Encompass_Preferred
Citric acid triammonium is formed by citric acid reacting with ammonia in a molar ratio of 1:3. It can be used as a carbon source to prepare carbon quantum dots (CDs). Higher nitrogen components in citric acid triammonium may promote nitrogen-based functional groups in CDs, leading to more efficient emission-color tunability.
  • Used as a carbon source for carbon quantum dots
  • Promotes nitrogen-based functional groups in CDs
  • Leads to efficient emission-color tunability
  • Human endogenous metabolite
Catalog No. 50-004-11197 Supplier Medchemexpress LLC Supplier No. HYB1529A50G
Citric acid triammonium is formed by citric acid reacting with ammonia in a molar ratio of 1:3. It can be used as a carbon source to prepare carbon quantum dots (CDs). Higher nitrogen components in citric acid triammonium may promote nitrogen-based functional groups in CDs, leading to more efficient emission-color tunability.
  • Used as a carbon source for carbon quantum dots
  • Promotes nitrogen-based functional groups in CDs
  • Leads to efficient emission-color tunability
  • Human endogenous metabolite