Learn More
Medchemexpress LLC Citric acid (tripotassium hydrate) | 6100-05-6 | 98.0% | 324.41 | 1 ML
SDP
Catalog No. 5000453403
Encompass_Preferred
Citric acid tripotassium hydrate is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. This compound can also cause oxidative damage to the liver by decreasing antioxidative enzyme activities. Additionally, it functions as an acidulant, emulsifier, sequestrant, and buffering agent across various industries.
  • Natural preservative and food tartness enhancer.
  • Induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells.
  • Causes oxidative damage of the liver by decreasing antioxidative enzyme activities.
  • Acts as an acidulant, emulsifier, sequestrant, and buffering agent.
Catalog No. 50-004-53403 Supplier Medchemexpress LLC Supplier No. HYW00915610MM/1ML
Citric acid tripotassium hydrate is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. This compound can also cause oxidative damage to the liver by decreasing antioxidative enzyme activities. Additionally, it functions as an acidulant, emulsifier, sequestrant, and buffering agent across various industries.
  • Natural preservative and food tartness enhancer.
  • Induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells.
  • Causes oxidative damage of the liver by decreasing antioxidative enzyme activities.
  • Acts as an acidulant, emulsifier, sequestrant, and buffering agent.