Learn More
Medchemexpress LLC Citric acid (tripotassium hydrate) | 6100-05-6 | 98.0% | 324.41 | 5 G
SDP
Catalog No. 5000366864
Encompass_Preferred
Citric acid tripotassium hydrate (Potassium citrate monohydrate) is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M and S phases in HaCaT cells. This compound can cause oxidative damage to the liver by decreasing antioxidative enzyme activities. It also serves as an acidulant, emulsifier, sequestrant, and buffering agent with wide applications across various industries. It is intended for research use only.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest in HaCaT cells
  • Acidulant, emulsifier, sequestrant, and buffering agent
  • Available as a solid, white to off-white in color
  • Soluble in water (120 mg/mL)
Catalog No. 50-003-66864 Supplier Medchemexpress LLC Supplier No. HYW0091565G
Citric acid tripotassium hydrate (Potassium citrate monohydrate) is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M and S phases in HaCaT cells. This compound can cause oxidative damage to the liver by decreasing antioxidative enzyme activities. It also serves as an acidulant, emulsifier, sequestrant, and buffering agent with wide applications across various industries. It is intended for research use only.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest in HaCaT cells
  • Acidulant, emulsifier, sequestrant, and buffering agent
  • Available as a solid, white to off-white in color
  • Soluble in water (120 mg/mL)