Learn More
Medchemexpress LLC Citric acid (tripotassium hydrate) | 6100-05-6 | 99.9% | 324.41 | 50 MG
SDP
Catalog No. 5000453529
Encompass_Preferred
Citric acid tripotassium hydrate (Standard) is the analytical standard of Citric acid tripotassium hydrate. It is intended for research and analytical applications, functioning as a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M and S phase in HaCaT cells, and can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. Additionally, it is used as an acidulant, emulsifier, sequestrant, and buffering agent across various industries.
  • Serves as a reference standard supplied for assay
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS applications
Catalog No. 50-004-53529 Supplier Medchemexpress LLC Supplier No. HYW009156R50MG
Citric acid tripotassium hydrate (Standard) is the analytical standard of Citric acid tripotassium hydrate. It is intended for research and analytical applications, functioning as a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M and S phase in HaCaT cells, and can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. Additionally, it is used as an acidulant, emulsifier, sequestrant, and buffering agent across various industries.
  • Serves as a reference standard supplied for assay
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS applications