Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, trisodium salt dihydrate, 99%, pure

Catalog No. AC227130050
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227130050 5 g Glass Bottle
AC227135000 500 g Plastic Bottle
AC227130010 1 kg Plastic Bottle
AC227130051 5 kg Plastic Bottle
AC227130250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC227130050 Supplier Thermo Scientific Chemicals Supplier No. 227130050
Add to Cart

CAS: 1545801 | C6H9Na3O9 | 294.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1545801
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.1
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass Bottle
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Quantity 5 g
Merck Index 15, 8737
Solubility Information Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 294.09
Percent Purity 0.99
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Citric acid, trisodium salt dihydrate
Show More Show Less
Hazard Category Signal Word: CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health:

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.