Learn More
Medchemexpress LLC Citric acid trisodium (sodium citrate) | 68-04-2 | 98.0% | 500 MG
SDP
Catalog No. 5000410295
Encompass_Preferred
Citric acid trisodium (Sodium citrate) is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid trisodium can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent used across many industries.
  • Natural preservative and food tartness enhancer.
  • Induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells.
  • Causes oxidative damage of the liver by decreasing antioxidative enzyme activities.
  • Functions as an acidulant, emulsifier, sequestrant, and buffering agent.
Catalog No. 50-004-10295 Supplier Medchemexpress LLC Supplier No. HYB2201500MG
Citric acid trisodium (Sodium citrate) is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid trisodium can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent used across many industries.
  • Natural preservative and food tartness enhancer.
  • Induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells.
  • Causes oxidative damage of the liver by decreasing antioxidative enzyme activities.
  • Functions as an acidulant, emulsifier, sequestrant, and buffering agent.