Learn More
Medchemexpress LLC Citric acid trisodium (Sodium citrate) | 68-04-2 | 99.95% | 10 G
SDP
Catalog No. 5000412307
Encompass_Preferred
Citric acid trisodium (Sodium citrate) is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells, and can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. Additionally, it serves as an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells
  • Acidulant
  • Emulsifier
  • Sequestrant
  • Buffering agent
Catalog No. 50-004-12307 Supplier Medchemexpress LLC Supplier No. HYB220110G
Citric acid trisodium (Sodium citrate) is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells, and can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. Additionally, it serves as an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells
  • Acidulant
  • Emulsifier
  • Sequestrant
  • Buffering agent