Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric/Phosphate buffer, pH 3.0, Ricca Chemical
SDP

Catalog No. p-7215415 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21138004A 4 L
R211380010F 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R21138004A Supplier Ricca Chemical Company Supplier No. R21138004A
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Citric/Phosphate Buffer
CAS 7732-18-5, 5949-29-1, 7558-79-4
CAS Max % 97.77, 1.65, 0.58
Color Clear, colorless
Concentration 1X
For Use With (Application) Water Analysis
pH 3
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Natural Poly Bottle
Specific Gravity 1.005-1.025
Sterility Non-sterile
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.