Learn More
Penetone Corporation Citrikleen | 5g Pail
Supplier:  Penetone Corporation 185028
Encompass_Preferred
Citrikleen is a patented, biodegradable, water-soluble, heavy-duty cleaner/degreaser formulated with an non-petroleum organic solvent and multicomponent surfactant emulsifier system to achieve superior cleaning and degreasing capabilities. It is used in concentrated form or at various water dilution rates. Citrikleen will rapidly penetrate and lift the widest range of petroleum, animal and vegetable based oils, fats and greases, which are then easily removed by water rinsing. The oilycontaminants in used Citrikleen solutions, left in a still tank or holding pond after cleaning, will rise to the surface and separate. This oil may be removed by top skimming. The remaining bottom layer is a biodegradable, reusable liquid cleaning solution which can be easily disposed of in accordance with local and/or governmental regulations if no longer needed
Catalog No. NC1788022