Learn More
AdipoGen Citrinalin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20177M001
Encompass_Preferred
Chemical. CAS: 1256635-19-4. Formula: C25H31N3O5. MW: 453.5. Isolated from fungus Penicillium sp. Antibiotic. Mycotoxin. Showed no antimicrobial, antifungal or cytotoxic activity in initial tests, therefore the acitivty profile of this compound is not known so far. Natural products that contain aliphatic nitro groups are extremely rare. In addition citrinalin A possesses three nitrogen atoms in chemically distinct environments which is rather unusual and therefore an attractive target for synthesis. Pseudo-enantiomer of Cyclopiamine A.
Catalog No. 50-205-3565