Learn More
Cayman Chemical Citrinin-13C13, Purity: ≥98%, 1.2 mL
SDP
Catalog No. 502989181
Encompass_Preferred
An internal standard for the quantification of citrinin by GC- or LC-MS
Catalog No. 50-298-9181 Supplier Cayman Chemical Supplier No. 312961.2 ML
An internal standard for the quantification of citrinin by GC- or LC-MS