Learn More
Medchemexpress LLC HY-N6746 1mg Medchemexpress, Citrinin CAS:518-75-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67461MG
Encompass_Preferred
Medchemexpress, HY-N6746 1mg Citrinin CAS:518-75-2 Citrinin is a mycotoxin which causes contamination in the food and is associated with different toxic effects. Citrinin is usually found together with another nephrotoxic mycotoxin, Ochratoxin A. Citrinin is also reported to possess a broad spectrum of bioactivities, including antibacterial, antifungal, and potential anticancer and neuro-protective effects in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2961