Learn More
TILLEY DISTRIBUTION INC CITRISOL 503 AC SOL 555LBDRUM
Catalog No. NC9893037
Encompass
ADM Citrosol 500 products are industrial grade 50% aqueous citric acid solutions (anhydrous basis), suitable for use in industrial applications which do not require USP or FCC grade citric acid.
Catalog No. NC9893037 Supplier TILLEY DISTRIBUTION INC Supplier No. CAL557
ADM Citrosol 500 products are industrial grade 50% aqueous citric acid solutions (anhydrous basis), suitable for use in industrial applications which do not require USP or FCC grade citric acid.