Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decon™ CitriSolv™ Hybrid Solvent and Clearing Agent
GSA_VA

Catalog No. 04355121 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-355-121 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-355-121 Supplier Decon™ Supplier No. 1601H

missing translation for 'validMainframeMsg'

Can be used as an alternative to xylene and ethyl acetate. CitriSolv Hybrid is soluble with absolute alcohol and will dissolve paraffin, glue, adhesives and mounting media.

  • A mixture of d-limonene and hydrocarbon material
  • Reduced citrus aroma
  • Increased flash point
  • Alternative to xylene

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Physical Form Liquid
Chemical Name or Material CitriSolv™Hybrid Solvent and Clearing Agent
CAS 5989-27-5, 25321-09-9, 128-37-0
Color Clear
Quantity 1 gal.
Molecular Weight (g/mol) 46.07 g/mol
Percent Purity 70%
Grade Laboratory
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.