Learn More
Stellar Solutions Inc CitriSurf 2310 Spray Gel | 22oz
SDP
Catalog No. NC1731531
Encompass
This item has a minimum order quantity of 12 per supplier requirements
CitriSurf 2310 is a high quality blend of chemicals formulated for cleaning, rust removal and passivation of stainless steel products. It is provided in a high viscosity form for easy application on weld joints, vertical surfaces, and other large items or areas that cannot be conveniently dipped in a bath. CitriSurf 2310 is specifically designed to provide low cost and efficient removal of all rust and free iron from the surface of stainless steel and other metals, providing the highest chrome oxide levels possible.
Catalog No. NC1731531 Supplier Stellar Solutions Inc Supplier No. CMP122310
This item has a minimum order quantity of 12 per supplier requirements
CitriSurf 2310 is a high quality blend of chemicals formulated for cleaning, rust removal and passivation of stainless steel products. It is provided in a high viscosity form for easy application on weld joints, vertical surfaces, and other large items or areas that cannot be conveniently dipped in a bath. CitriSurf 2310 is specifically designed to provide low cost and efficient removal of all rust and free iron from the surface of stainless steel and other metals, providing the highest chrome oxide levels possible.