Learn More
ATCC Citrobacter freundii
Catalog No. 502383634
Encompass
Type strain. Reference strain. Quality control strain for Sensititre products. Produces restriction endonuclease CfrAI.
Catalog No. 50-238-3634 Supplier ATCC Supplier No. 8090
Type strain. Reference strain. Quality control strain for Sensititre products. Produces restriction endonuclease CfrAI.