Learn More
ATCC Citrobacter rodentium; Strain DBS100
Catalog No. 502383166
Encompass
Biotype 4280; causes murine colonic hyperplasia.
Catalog No. 50-238-3166 Supplier ATCC Supplier No. 51459
Biotype 4280; causes murine colonic hyperplasia.