Learn More
ATCC Salmonella enterica; subsp. diarizonae
Supplier:  ATCC 29934
Encompass
Formerly Salmonella choleraesuis subsp. diarizonae. Control strain for detection of Salmonella. DNA hybridization reference strain.
Catalog No. 50-238-0485