Learn More
Medchemexpress LLC Citronellol | 106-22-9 | 98.4% | 156.27 | 25 G
SDP
Catalog No. 5000366440
Encompass_Preferred
Citronellol is an orally active inducer of apoptosis. It prevents oxidative stress, mitochondrial dysfunction, and apoptosis in the SH-SY5Y cell Parkinson's disease model. It can also induce necroptosis in human lung cancer cells through the TNF-α pathway and accumulation of ROS.
  • Regulates autophagy pathway by reducing LC-3 and p62 levels.
  • Inhibits oxidative stress and neuroinflammation.
  • Provides neuroprotective effects in Parkinson's models.
  • Exhibits anti-fungal activity by inhibiting ergosterol synthesis.
  • Improves cell viability in neuroblastoma cells.
  • Offers cardioprotective effects in myocardial ischemia models.
  • Provides kidney protection in rhabdomyolysis models.
  • Demonstrates anti-tumor activity in NSCLC models.
Catalog No. 50-003-66440 Supplier Medchemexpress LLC Supplier No. 25GHYW010201
Citronellol is an orally active inducer of apoptosis. It prevents oxidative stress, mitochondrial dysfunction, and apoptosis in the SH-SY5Y cell Parkinson's disease model. It can also induce necroptosis in human lung cancer cells through the TNF-α pathway and accumulation of ROS.
  • Regulates autophagy pathway by reducing LC-3 and p62 levels.
  • Inhibits oxidative stress and neuroinflammation.
  • Provides neuroprotective effects in Parkinson's models.
  • Exhibits anti-fungal activity by inhibiting ergosterol synthesis.
  • Improves cell viability in neuroblastoma cells.
  • Offers cardioprotective effects in myocardial ischemia models.
  • Provides kidney protection in rhabdomyolysis models.
  • Demonstrates anti-tumor activity in NSCLC models.