Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citronellyl acetate | 150-84-5 | MFCD00015039 | 1 G
SDP

Catalog No. 5000417970
Encompass_Preferred

Citronellyl acetate is a monoterpene product derived from the secondary metabolism of plants. It functions as an orally active antinociceptive agent, modulating various receptors and kinases involved in nociception, and is suitable for acute pain research.

  • Derived from secondary metabolism of plants
  • Functions as an orally active antinociceptive agent
  • Modulates TRPV1, TRPM8, ASIC, glutamate receptors, PKC, and PKA-mediated nociception
  • Suitable for acute pain research
  • Purity of 98.0%
  • Appearance as a colorless to light yellow liquid

Catalog No. 50-004-17970 Supplier Medchemexpress LLC Supplier No. HYN7144A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citronellyl acetate is a monoterpene product derived from the secondary metabolism of plants. It functions as an orally active antinociceptive agent, modulating various receptors and kinases involved in nociception, and is suitable for acute pain research.

  • Derived from secondary metabolism of plants
  • Functions as an orally active antinociceptive agent
  • Modulates TRPV1, TRPM8, ASIC, glutamate receptors, PKC, and PKA-mediated nociception
  • Suitable for acute pain research
  • Purity of 98.0%
  • Appearance as a colorless to light yellow liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.