Learn More
Cayman Chemical (+)-β-Citronellol, Purity: ≥95%, CAS: 1117-61-9, 250 mg
SDP
Catalog No. 502995337
Encompass_Preferred
A monoterpene; inhibits degranulation of cultured mast cells by 21.3% at 0.5 mM; does not reduce the perceived bitterness of caffeine at 5.9 nM
Catalog No. 50-299-5337 Supplier Cayman Chemical Supplier No. 23464250 MG
A monoterpene; inhibits degranulation of cultured mast cells by 21.3% at 0.5 mM; does not reduce the perceived bitterness of caffeine at 5.9 nM