Learn More
Cayman Chemical CItrulInATd CATnInhuma 100ug
SDP
Catalog No. 502989232
Encompass_Preferred
Source: Recombinant N-terminal His-tagged human β-catenin expressed in E. coli, citrullinated by PAD2 • Amino acids: 138-781 • MW: 72 kDa
Catalog No. 50-298-9232 Supplier Cayman Chemical Supplier No. 29921100 UG
Source: Recombinant N-terminal His-tagged human β-catenin expressed in E. coli, citrullinated by PAD2 • Amino acids: 138-781 • MW: 72 kDa