Learn More
Cayman Chemical CItrulInATd GFAPR416 Mon 300ug
SDP
Catalog No. 502940002
Encompass_Preferred
Immunogen: A peptide from the C-terminal region of human GFAP citrullinated at R416 • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) Citrullinated GFAP (-) Native GFAP, other citrullinated proteins • Applications: ELISA, IHC, WB • MW: 37 kDa
Catalog No. 50-294-0002 Supplier Cayman Chemical Supplier No. 27795300 UG
Immunogen: A peptide from the C-terminal region of human GFAP citrullinated at R416 • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) Citrullinated GFAP (-) Native GFAP, other citrullinated proteins • Applications: ELISA, IHC, WB • MW: 37 kDa