Learn More
Cayman Chemical CItrulInATd GRP78humn re 100ug
SDP
Catalog No. 502932645
Encompass_Preferred
Source: N-Terminal His-tagged human GRP78 purified from E. coli, citrullinated by PAD2 • MW: 74.6 kDa
Catalog No. 50-293-2645 Supplier Cayman Chemical Supplier No. 25107100 UG
Source: N-Terminal His-tagged human GRP78 purified from E. coli, citrullinated by PAD2 • MW: 74.6 kDa