Learn More
Cayman Chemical CItrulInATd HIston H2Bhu 100ug
SDP
Catalog No. 502990551
Encompass_Preferred
Source: Recombinant human histone H2B type 1-A expressed in E. coli, citrullinated with human recombinant PAD4 • Amino acids: 2-127 (full length) • MW: 14.04 kDa
Catalog No. 50-299-0551 Supplier Cayman Chemical Supplier No. 30133100 UG
Source: Recombinant human histone H2B type 1-A expressed in E. coli, citrullinated with human recombinant PAD4 • Amino acids: 2-127 (full length) • MW: 14.04 kDa