Learn More
Cayman Chemical CItrulInATd HIston H4hum 100ug
Supplier: Cayman Chemical 17927100 UG
Source: Recombinant protein expressed in E. coli, citrullinated with human recombinant PAD4 • Amino acids: 1-103 (full-length) • MW: 11.5 kDa