Learn More
Cayman Chemical CItrulInATd HIston H4hum 100ug
SDP
Catalog No. 502916010
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged histone H4 expressed in E. coli, citrullinated with recombinant human PAD4 • Amino acids: 1-103 (full length) • MW: 13.4 kDa
Catalog No. 50-291-6010 Supplier Cayman Chemical Supplier No. 38258100 UG
Source: Recombinant human N-terminal His-tagged histone H4 expressed in E. coli, citrullinated with recombinant human PAD4 • Amino acids: 1-103 (full length) • MW: 13.4 kDa