Learn More
Cayman Chemical CItrulInATd VImentInhumn 100ug
SDP
Catalog No. 502986576
Encompass_Preferred
Source: Human recombinant vimentin purified from E. coli • MW: 53.5 kDa
Catalog No. 50-298-6576 Supplier Cayman Chemical Supplier No. 21942100 UG
Source: Human recombinant vimentin purified from E. coli • MW: 53.5 kDa