Learn More
Cayman Chemical CItrulInated Colagen TYP I
SDP
Supplier:  Cayman Chemical 39297200 UG
Encompass_Preferred
Immunogen: Bovine collagen type IIA • Host: Chimeric monoclonal antibody • Species Reactivity: (+) Human, mouse, bovine • Cross Reactivity: (+) Citrullinated form of the triple helical collagen type II C1 epitope (amino acids 359-369), denatured collagen type II, denatured collagen type II triple helical C1 epitope (amino acids 359-369). Some cross reactivity to non-citrullinated epitopes on collagen type II • Applications: ELISA, IHC
Catalog No. 50-291-7234